1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-4-bromo-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulphonic Acid Sodium Salt, Bromaminic acid Sodium Salt, Bromaminic acid sodium salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate

A944088-100G A944088-25G A944088-5G
77200-358EA 105.06 USD
77200-358 77200-360 77200-362
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt 98%
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt

Precaución: for Research Use Only

Formula: C₁₄H₈BrNO₅S
MW: 404.17 g/mol
MDL Number: MFCD00019160
CAS Number: 6258-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR