1-Benzoylpiperazine hydrochloride 98%

Supplier: Ambeed
Warning

A942163-1G A942163-25G A942163-5G
77200-352EA 10.3 USD
77200-352 77200-354 77200-356
1-Benzoylpiperazine hydrochloride 98%
1-Benzoylpiperazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O
MW: 226.70 g/mol
MDL Number: MFCD00459578
CAS Number: 56227-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR