4-Bromo-3-fluoro-2-methylaniline 97%

Supplier: Ambeed
Warning

A941577-1G A941577-10G A941577-25G A941577-5G
77171-986EA 20 USD
77171-986 77614-658 77893-746 77893-747
4-Bromo-3-fluoro-2-methylaniline 97%
4-Bromo-3-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD09800761
CAS Number: 127408-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR