[3-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(1H-pyrazol-1-yl)phenyl)boronic acid

A940384-100MG A940384-250MG A940384-5G A940384-1G
77171-980EA 42.99 USD
77171-980 77171-982 77171-984 77893-743
[3-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%
[3-(1H-Pyrazol-1-yl)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₂
MDL Number: MFCD05864582
CAS Number: 476620-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR