5-Bromo-1-chloro-2-methyl-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A935098-1G A935098-250MG
77171-964EA 134 USD
77171-964 77893-740
5-Bromo-1-chloro-2-methyl-3-nitrobenzene 97%
5-Bromo-1-chloro-2-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MDL Number: MFCD07781356
CAS Number: 885519-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR