2-(2-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methylthiophenylboronic acid pinacol ester

A935013-100MG A935013-1G A935013-250MG
77200-330EA 18 USD
77200-330 77200-332 77200-334
2-(2-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(2-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
MDL Number: MFCD29088253
CAS Number: 1436431-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR