1,1'-Dimethylferrocene 98%

Supplier: Ambeed
Warning

A933793-1G A933793-25G A933793-5G
77200-324EA 18 USD
77200-324 77200-326 77200-328
1,1'-Dimethylferrocene 98%
1,1'-Dimethylferrocene

Precaución: for Research Use Only

Formula: C₁₂H₁₄Fe
MDL Number: MFCD00001425
CAS Number: 1291-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR