2-Cyanomethyl benzoic acid ethyl ester 98+%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-(cyanomethyl)benzoate

A932903-1G A932903-25G A932903-5G
77171-958EA 25.75 USD
77171-958 77171-960 77171-962
2-Cyanomethyl benzoic acid ethyl ester 98+%
2-Cyanomethyl benzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
MDL Number: MFCD00062509
CAS Number: 19821-21-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR