m-Toluic hydrazide 98%

Supplier: Ambeed
Warning

Synonyms: m-Toluic acid hydrazide, 3-Methylbenzoic hydrazide

A932733-10G A932733-1G A932733-5G
77200-318EA 19 USD
77200-318 77200-320 77200-322
m-Toluic hydrazide 98%
m-Toluic hydrazide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD00014760
CAS Number: 13050-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR