Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
C34, TLR4-IN-C34, (2R,3S,4R,5R,6S)-5-Acetamido-2-(acetoxymethyl)-6-isopropoxytetrahydro-2H-pyran-3,4-diyl diacetate, Isopropyl 3,4,6-tri-O-acetyl-α-D-GlcNAc
A932319-10MG
A932319-1MG
A932319-25MG
A932319-5MG
77171-950EA
105
USD
77171-950
77171-952
77171-954
77171-956
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside 98%
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside
Precaución: for Research Use Only
Formula:
C₁₇H₂₇NO₉ |
MDL Number:
MFCD28053519 CAS Number: 40592-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...