3-Fluorothiophenol 97%

Supplier: Ambeed
Danger

A927679-1G A927679-5G A927679-25G
77171-938EA 13 USD
77171-938 77171-940 77893-734
3-Fluorothiophenol 97%
3-Fluorothiophenol

Precaución: for Research Use Only

Formula: C₆H₅FS
MDL Number: MFCD00040227
CAS Number: 2557-77-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR