8-Oxa-2-azaspiro[4.5]decane hydrochloride 98%

Supplier: Ambeed
Warning

A927428-1G A927428-5G A927428-250MG
77171-934EA 33 USD
77171-934 77171-936 77893-733
8-Oxa-2-azaspiro[4.5]decane hydrochloride 98%
8-Oxa-2-azaspiro[4.5]decane hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO·ClH
MDL Number: MFCD18325189
CAS Number: 1408074-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR