N-(1,3-Diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide 97%

Supplier: Ambeed
Warning

Synonyms: VU-29

A927285-10MG A927285-1MG A927285-25MG A927285-50MG A927285-5MG
77200-308EA 40.17 USD
77200-308 77200-310 77200-312 77200-314 77200-316
N-(1,3-Diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide 97%
N-(1,3-Diphenyl-1H-pyrazol-5-yl)-4-nitrobenzamide

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₄O₃
MW: 384.39 g/mol
MDL Number: MFCD12546140
CAS Number: 890764-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR