1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Isopropyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)boronic acid

A926879-1G A926879-250MG
77171-930EA 166.86 USD
77171-930 77171-932
1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid 98%
1-Isopropyl-3-(trifluoromethyl)pyrazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BF₃N₂O₂
MW: 221.973 g/mol
MDL Number: MFCD24448764
CAS Number: 1361380-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR