10-Methyl-9(10H)-acridone 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-9-acridone, 10-Methylacridin-9(10H)-one

A925319-1G A925319-250MG A925319-5G A925319-25G
77200-302EA 15 USD
77200-302 77200-304 77200-306 77747-687
10-Methyl-9(10H)-acridone 98%
10-Methyl-9(10H)-acridone

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MDL Number: MFCD00005024
CAS Number: 719-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR