Methyl 2-amino-5-iodo-4-methoxybenzoate 96%

Supplier: Ambeed
Warning

A922373-100MG A922373-1G A922373-250MG
77200-296EA 45.32 USD
77200-296 77200-298 77200-300
Methyl 2-amino-5-iodo-4-methoxybenzoate 96%
Methyl 2-amino-5-iodo-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀INO₃
MW: 307.08 g/mol
MDL Number: MFCD24387259
CAS Number: 1256958-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR