2,5-Diiodo-1-methylimidazole 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Diiodo-1-methyl-1H-imidazole

A922249-100MG A922249-1G
77171-928EA 25 USD
77171-928 77595-492
2,5-Diiodo-1-methylimidazole 98%
2,5-Diiodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₄I₂N₂
MW: 333.9
MDL Number: MFCD02179540
CAS Number: 86026-81-5
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR