1-Methyl-1H-1,2,3-triazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A922230-1G A922230-25G A922230-5G
77171-922EA 14 USD
77171-922 77171-924 77171-926
1-Methyl-1H-1,2,3-triazole-4-carbaldehyde 98%
1-Methyl-1H-1,2,3-triazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MDL Number: MFCD10696345
CAS Number: 16681-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR