1,9-Bis(acryloyloxy)nonane 98% stabilized

Supplier: Ambeed
Warning

Synonyms: Nonanediol Diacrylate, Nonamethylene Glycol Diacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A921928-1000G A921928-100G A921928-500G
77200-290EA 167.89 USD
77200-290 77200-292 77200-294
1,9-Bis(acryloyloxy)nonane 98% stabilized
1,9-Bis(acryloyloxy)nonane

Precaución: for Research Use Only

Formula: C₁₅H₂₄O₄
MW: 268.35 g/mol
MDL Number: MFCD00080592
CAS Number: 107481-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR