1-(5-Isopropoxypyridin-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A921293-100MG A921293-1G A921293-250MG
77171-916EA 50.47 USD
77171-916 77171-918 77171-920
1-(5-Isopropoxypyridin-2-yl)ethanone 95%
1-(5-Isopropoxypyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
CAS Number: 1198166-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR