PF-04418948 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(4-fluorobenzoyl)-3-[(6-methoxynaphthalen-2-yl)oxymethyl]azetidine-3-carboxylic acid, 1-(4-Fluorobenzoyl)-3-(((6-methoxynaphthalen-2-yl)oxy)methyl)azetidine-3-carboxylic acid

A920639-10MG A920639-1G A920639-1MG A920639-25MG A920639-50MG A920639-5MG A920639-250MG A920639-100MG
77171-904EA 69 USD
77171-904 77171-906 77171-908 77171-910 77171-912 77171-914 77893-726 77893-725
PF-04418948 98+%
PF-04418948

Precaución: for Research Use Only

Formula: C₂₃H₂₀FNO₅
MW: 409.41 g/mol
MDL Number: MFCD24387541
CAS Number: 1078166-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR