4-(4-Chlorophenyl)-3-thiosemicarbazide 98+%

Supplier: Ambeed
Danger

Synonyms: N-(4-Chlorophenyl)hydrazinecarbothioamide

A919882-1G A919882-250MG A919882-5G
77200-278EA 14.42 USD
77200-278 77200-280 77200-282
4-(4-Chlorophenyl)-3-thiosemicarbazide 98+%
4-(4-Chlorophenyl)-3-thiosemicarbazide

Precaución: for Research Use Only

Formula: C₇H₈ClN₃S
MW: 201.68 g/mol
MDL Number: MFCD00041295
CAS Number: 22814-92-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR