2-Bromo-4-chloro-1,3-difluorobenzene 98%

Supplier: Ambeed
Warning

A918789-25G A918789-5G
77200-274EA 265 USD
77200-274 77200-276
2-Bromo-4-chloro-1,3-difluorobenzene 98%
2-Bromo-4-chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD12922593
CAS Number: 229180-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR