(2-Bromothiazol-5-yl)methanamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A918743-100MG
A918743-250MG
77171-900EA
199.82
USD
77171-900
77171-902
(2-Bromothiazol-5-yl)methanamine hydrochloride 95%
(2-Bromothiazol-5-yl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₆BrClN₂S MW: 229.52 g/mol |
MDL Number:
MFCD08143030 CAS Number: 1001413-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...