Pentamethyldisiloxane ≥95% (by GC)

Supplier: Ambeed
Danger

Synonyms: 1,1,1,3,3-Pentamethyldisiloxane

A917895-5G A917895-25G
77171-898EA 17.51 USD
77171-898 77569-636
Pentamethyldisiloxane ≥95% (by GC)
Pentamethyldisiloxane

Precaución: For research use only.

Formula: C₅H₁₆OSi₂
MW: 148.35 g/mol
MDL Number: MFCD00053658
CAS Number: 1438-82-0
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR