1-(Benzo[d][1,3]dioxol-5-yl)pentan-1-one 95%

Supplier: Ambeed
Warning

A916493-1G A916493-250MG A916493-5G
77200-268EA 58 USD
77200-268 77200-270 77200-272
1-(Benzo[d][1,3]dioxol-5-yl)pentan-1-one 95%
1-(Benzo[d][1,3]dioxol-5-yl)pentan-1-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MDL Number: MFCD09998839
CAS Number: 63740-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR