1,3-Dicyclohexylbenzimidazolium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dicyclohexyl-1H-benzo[d]imidazol-3-ium chloride, 1,3-Dicyclohexyl-1H-benzimidazol-3-ium chloride

A915953-100MG A915953-1G
77200-264EA 22 USD
77200-264 77200-266
1,3-Dicyclohexylbenzimidazolium chloride 98%
1,3-Dicyclohexylbenzimidazolium chloride

Precaución: for Research Use Only

Formula: C₁₉H₂₇ClN₂
MW: 318.89 g/mol
MDL Number: MFCD08705248
CAS Number: 1034449-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR