3,4-Dimethoxybenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Veratroyl chloride, 3,4-Dimethoxybenzene-1-carbonyl chloride

A914059-1G A914059-25G A914059-5G
77171-890EA 12 USD
77171-890 77171-892 77552-706
3,4-Dimethoxybenzoyl chloride 98%
3,4-Dimethoxybenzoyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD00000674
CAS Number: 3535-37-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR