1-Benzyl-3-methylimidazolium tetrafluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

A913528-100G A913528-1G A913528-25G A913528-5G
77200-256EA 74 USD
77200-256 77200-258 77200-260 77200-262
1-Benzyl-3-methylimidazolium tetrafluoroborate 98%
1-Benzyl-3-methylimidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BF₄N₂
MW: 260.04
MDL Number: MFCD07784449
CAS Number: 500996-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR