1,1,1,3,5,5,5-Heptamethyltrisiloxane 98%

Supplier: Ambeed
Danger

Synonyms: Methylbis(trimethylsilyloxy)silane

A913309-10G A913309-5G A913309-25G A913309-100G
77200-252EA 18 USD
77200-252 77200-254 77747-679 77747-678
1,1,1,3,5,5,5-Heptamethyltrisiloxane 98%
1,1,1,3,5,5,5-Heptamethyltrisiloxane

Precaución: for Research Use Only

Formula: C₇H₂₂O₂Si₃
MW: 222.51
MDL Number: MFCD00048000
CAS Number: 1873-88-7
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR