1-Chloro-3-(3-chloropropoxy)propane 98%

Supplier: Ambeed
Danger

A911774-100MG A911774-1G A911774-250MG A911774-5G
77171-876EA 11 USD
77171-876 77171-878 77171-880 77893-710
1-Chloro-3-(3-chloropropoxy)propane 98%
1-Chloro-3-(3-chloropropoxy)propane

Precaución: for Research Use Only

Formula: C₆H₁₂Cl₂O
MDL Number: MFCD08437454
CAS Number: 629-36-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR