1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene 97%

Supplier: Ambeed
Warning

Synonyms: OXD-7, 1,3-Bis(5-(4-(tert-butyl)phenyl)-1,3,4-oxadiazol-2-yl)benzene, 1,3-Bis[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]benzene

A911747-100MG A911747-250MG A911747-1G
77200-242EA 21.63 USD
77200-242 77200-244 77308-506
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene 97%
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene

Precaución: for Research Use Only

Formula: C₃₀H₃₀N₄O₂
MW: 478.60 g/mol
MDL Number: MFCD03093214
CAS Number: 138372-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR