2-Amino-2-(2,5-dichlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A904314-100MG A904314-1G A904314-250MG
77200-218EA 53.56 USD
77200-218 77200-220 77200-222
2-Amino-2-(2,5-dichlorophenyl)acetic acid 97%
2-Amino-2-(2,5-dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
MDL Number: MFCD02662395
CAS Number: 500695-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR