5-Fluoroindoline 95%

Supplier: Ambeed
Warning

A904024-1G A904024-250MG A904024-5G
77171-860EA 61.8 USD
77171-860 77171-862 77171-864
5-Fluoroindoline 95%
5-Fluoroindoline

Precaución: for Research Use Only

Formula: C₈H₈FN
MW: 137.16 g/mol
MDL Number: MFCD00214461
CAS Number: 2343-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR