1-Butyl-2,3-dimethylimidazolium bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-2,3-dimethyl-1H-imidazol-3-ium bromide

A899255-100G A899255-25G A899255-5G
77200-204EA 81.37 USD
77200-204 77200-206 77200-208
1-Butyl-2,3-dimethylimidazolium bromide 98%
1-Butyl-2,3-dimethylimidazolium bromide

Precaución: for Research Use Only

Formula: C₉H₁₇BrN₂
MW: 233.15 g/mol
MDL Number: MFCD03427615
CAS Number: 475575-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR