4-(Benzenesulfonyl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Phenylsulphonyl)benzaldehyde

A898728-100MG A898728-1G
77200-200EA 20 USD
77200-200 77200-202
4-(Benzenesulfonyl)benzaldehyde 97%
4-(Benzenesulfonyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃S
MDL Number: MFCD00444612
CAS Number: 66-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR