(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-N-Me-Lys(Boc)-OH
A897893-100MG
A897893-250MG
A897893-1G
A897893-5G
77171-834EA
24
USD
77171-834
77171-836
77595-440
77893-688
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid 95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid
Precaución: for Research Use Only
Formula:
C₂₇H₃₄N₂O₆ MW: 482.56 g/mol |
MDL Number:
MFCD08702737 CAS Number: 1793105-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...