1-(2-Methoxyethyl)-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A897719-100MG A897719-1G A897719-250MG
77200-194EA 23 USD
77200-194 77200-196 77200-198
1-(2-Methoxyethyl)-4-nitrobenzene 95%
1-(2-Methoxyethyl)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MDL Number: MFCD00634283
CAS Number: 69628-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR