3,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridazine 97%

Supplier: Ambeed
Warning

A897524-100MG A897524-1G A897524-250MG A897524-5G
77171-826EA 39 USD
77171-826 77171-828 77171-830 77171-832
3,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridazine 97%
3,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridazine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BCl₂N₂O₂
MDL Number: MFCD12827708
CAS Number: 919197-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR