Methyl 3-aminofuro[2,3-b]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

A895481-1G A895481-250MG
77200-190EA 101.97 USD
77200-190 77200-192
Methyl 3-aminofuro[2,3-b]pyridine-2-carboxylate 97%
Methyl 3-aminofuro[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.17 g/mol
MDL Number: MFCD12024763
CAS Number: 181283-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR