Tris(2,2'-bipyridine)nickel dichloride 98%

Supplier: Ambeed
Danger

A890246-5G A890246-1G A890246-25G
77171-808EA 91 USD
77171-808 77614-594 77614-596
Tris(2,2'-bipyridine)nickel dichloride 98%
Tris(2,2'-bipyridine)nickel dichloride

Precaución: for Research Use Only

Formula: C₃₀H₂₄Cl₂N₆Ni
CAS Number: 14751-84-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR