(S)-3-Benzylmorpholine 95%

Supplier: Ambeed
Warning

A889009-100MG A889009-1G A889009-250MG
77200-172EA 28.84 USD
77200-172 77200-174 77200-176
(S)-3-Benzylmorpholine 95%
(S)-3-Benzylmorpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.24 g/mol
MDL Number: MFCD11519115
CAS Number: 77897-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR