Pyren-1-ylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Pyren-1-ylmethanamine HCl

A888007-100MG A888007-250MG A888007-1G A888007-5G
77200-168EA 12 USD
77200-168 77200-170 77618-490 77618-492
Pyren-1-ylmethanamine hydrochloride 97%
Pyren-1-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₄ClN
MDL Number: MFCD00192425
CAS Number: 93324-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR