3-Fluoro-p-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoro-4-methylaniline, 4-Amino-2-fluorotoluene

A886940-10G A886940-25G A886940-5G
77200-162EA 12.36 USD
77200-162 77200-164 77200-166
3-Fluoro-p-toluidine 98%
3-Fluoro-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD00007762
CAS Number: 452-77-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR