Ethyl 2-((tert-butoxycarbonyl)amino)-3-nitrobenzoate 95%

Supplier: Ambeed
Warning

A884767-100G A884767-1G A884767-25G A884767-500G A884767-5G
77171-796EA 11 USD
77171-796 77171-798 77171-800 77171-802 77171-804
Ethyl 2-((tert-butoxycarbonyl)amino)-3-nitrobenzoate 95%
Ethyl 2-((tert-butoxycarbonyl)amino)-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₆
MDL Number: MFCD09833364
CAS Number: 136285-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR