4-Bromo-2-methylbenzo[d]oxazole 97%

Supplier: Ambeed
Warning

A884709-100MG A884709-1G A884709-250MG
77171-790EA 150.38 USD
77171-790 77171-792 77171-794
4-Bromo-2-methylbenzo[d]oxazole 97%
4-Bromo-2-methylbenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.04 g/mol
MDL Number: MFCD25978059
CAS Number: 217326-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR