2-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A883681-1G A883681-250MG A883681-5G
77171-784EA 59.74 USD
77171-784 77171-786 77171-788
2-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
2-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₅
MW: 277.08 g/mol
MDL Number: MFCD22493666
CAS Number: 1184259-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR