4-(2,4-Difluorophenoxy)piperidine 97%

Supplier: Ambeed
Warning

A882072-1G A882072-250MG A882072-25G A882072-5G
77171-768EA 37.08 USD
77171-768 77171-770 77171-772 77684-846
4-(2,4-Difluorophenoxy)piperidine 97%
4-(2,4-Difluorophenoxy)piperidine

Precaución: For research use only.

Formula: C₁₁H₁₃F₂NO
MW: 213.22 g/mol
MDL Number: MFCD08687571
CAS Number: 367501-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR