N,N-Dimethyl-2-(3,4,5-trifluorophenoxy)ethanamine 97%

Supplier: Ambeed
Danger

A880374-250MG A880374-5G
77171-764EA 19.57 USD
77171-764 77171-766
N,N-Dimethyl-2-(3,4,5-trifluorophenoxy)ethanamine 97%
N,N-Dimethyl-2-(3,4,5-trifluorophenoxy)ethanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₃NO
MW: 219.2 g/mol
MDL Number: MFCD28506167
CAS Number: 1708924-46-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR