(S)-tert-Butyl 3-((methylsulfonyl)oxy)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A878895-1G A878895-250MG
77171-760EA 22 USD
77171-760 77171-762
(S)-tert-Butyl 3-((methylsulfonyl)oxy)piperidine-1-carboxylate 95%
(S)-tert-Butyl 3-((methylsulfonyl)oxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₅S
MDL Number: MFCD09953027
CAS Number: 940890-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR